Tweezers, Straight Fine Tips, Boley Style, Excelta Corp®
Supplier: Excelta
AA AA-S AA-SA AA-SA-PI AA-SA-SE AA-SE AA-S-SE MM MM-SA-SE MM-SE AA-SA-PI-ET GG-SA
63043-198EA 91.48 CAD
63043-198 63043-202 63043-204 63043-208 63043-210 63043-212 63043-214 63043-480 63043-482 63043-484 89411-710 89411-752 89411-768 89411-822 89411-866
Tweezers, Straight Fine Tips, Boley Style, Excelta Corp®
Forceps
Designed for general assembly.
  • Strong tips for general assembly use
  • Available in different materials for different applications
  • Three different quality grades available
  • Option of ergonomic grips
These 'BOLEY' straight medium point general purpose tweezer are offered in 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat. The 'PH' suffix denotes plastic coated handles.