J.T.Baker® BAKERBOND® Poly Hi-Propyl™, Mixed Mode Hydrophobic Interaction Chromatography (HIC) Resin, Lab Column
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR6064-07 BAKR6064-25
CAJT6064-07EA 601.35 CAD
CAJT6064-07 CAJT6064-25
J.T.Baker® BAKERBOND® Poly Hi-Propyl™, Mixed Mode Hydrophobic Interaction Chromatography (HIC) Resin, Lab Column
Chromatography Columns
The J.T.Baker® BAKERBOND® Poly Hi-Propyl™ mixed mode hydrophobic interaction chromatography (HIC) resin provides improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining hydrophobic functionality with weak anionic exchange sites, BAKERBOND® Poly Hi-Propyl™ mixed mode HIC media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® Poly Hi-Propyl™ mixed mode HIC resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Ordering information: Available in conveniently-packaged 5 packs, the 1 and 5 ml lab columns are designed for direct connection to many popular chromatography instruments including GE ÄKTA™ and Bio-Rad NGC™. For bulk sizes (50, 100, 500 ml, 1 and 5 L), see the Complementary Products tab on the ordering table.