J.T.Baker® BAKERBOND® PolyCSX™, Multimode Ion Exchange (IEX) Chromatography Resin, Lab Column
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR6061-07 BAKR6061-25
CAJT6061-07EA 601.35 CAD
CAJT6061-07 CAJT6061-25
J.T.Baker® BAKERBOND® PolyCSX™, Multimode Ion Exchange (IEX) Chromatography Resin, Lab Column
Chromatography Columns
The J.T.Baker® BAKERBOND® PolyCSX™ multimode ion exchange (IEX) chromatography resin functions as a strong cation exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong cation exchange functionality with weak cationic and anionic exchange sites, BAKERBOND® PolyCSX™ multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyCSX™ ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Ordering information: Available in conveniently-packaged five packs, the 1 and 5 ml lab columns are designed for direct connection to many popular chromatography instruments including GE ÄKTA™ and Bio-Rad NGC™. For bulk sizes (50, 100, 500, 1 and 5 L), see the Complementary Products tab on the ordering table.