Cutters, Standoff Shear, Large Frame, Excelta
Supplier: Excelta
903 907-1 907-42A 907-5 907-59 903A 903A-L
63043-012EA 1118.99 CAD
63043-012 63043-018 63043-032 63043-034 63043-036 94024-392 94024-394
Cutters, Standoff Shear, Large Frame, Excelta
Cutters
Reduces shock on delicate solder connections and sensitive components soldered into a printed circuit board.
  • Anti-shock trimming of leads to specific length
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
This popular LARGE HEAD anti-shock shear cutter ensures that each and every lead is cut precisely to the desire length. There are many different configurations of this tool. Those with 'CUSTOM' suffixes require the buyer to specificy the desired lead length. They feature an ergonomically cushioned antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. These tools are made of carbon tool steel and have flame hardened cutting edges.