VWR® Disposable Face Shields
Supplier: Avantor
VWR®
V2
76443-134CS 652.22 CAD
76443-134
VWR® Disposable Face Shields
Face Shields
Perfect for use in environments with light particle, splash, or spray, the VWR® Disposable Face Shield is a cost effective and reliable way to keep covered.
  • Material: APET
  • Anti-fog inner and outer coating
  • PE protective film
  • Comfortable PU foam for extended wear
  • Elastic band to maintain comfortable fit
Face shields should be worn with ANSI approved respiratory protection, as needed.
Packaging: Packaged 200 per case.