VWR® Surgical Facemasks with Earloops, ASTM 1, 2, and 3
Supplier: VWR International
76437-414 76437-416 76437-418
76437-414CS 159.39 CAD
76437-414 76437-416 76437-418
VWR® Surgical Facemasks with Earloops, ASTM 1, 2, and 3
Masks Surgical Masks
These ASTM rated surgical masks offer various protection levels to fit specific working environments.
  • Multiple layers
  • Comfortable earloops
  • Adjustable nosewire to provide snug fit
  • Inner film layer to maximize breathibility
  • Designed for fluid protection
  • Latex-free
These masks are designed to protect against various ranges of fluid, spray, or debris. When worn properly, they will help to minimze contamination caused by exhaled microorganisms and reduce potential exposure to particulates and fluids.
ASTM Level 1 masks provide a fluid resistance of 80More Product Information