SmartWall® Single Shelf Support
Supplier: Metropolitan Wire
SWS14K4 SWS18K4 SWS21K4 SWS24K4
76463-154EA 127.44 CAD
76463-154 76463-156 76463-158 76463-160
SmartWall® Single Shelf Support
Furniture Shelves
Create a unique storage and workstation system for often underutilized wall space.
  • Mount directly to slotted uprights
  • Available in lengths to hold 14", 18", 21" and 24" shelves
  • Single shelf supports are used on each end of a single unit or side-by-side unit
  • No tools are necessary to assemble
Suitable for use with Super Erecta®, MetroMax® Q, and MetroMax® i shelves.
Can be used with SmartWall® Uprights to create an easily adjustable wall shelving system.
Metroseal Epoxy finishes feature Microban® antimicrobial product protection which helps to inhibit the growth of microorganisms such as bacteria, mold and fungi which can cause odors, stains and product degradation.