Super Erecta® Direct Wall Mount Double Shelf Bracket
Supplier: Metropolitan Wire
2WD14K4 2WD18K4 2WD24K4
76463-088EA 177.61 CAD
76463-088 76463-090 76463-092
Super Erecta® Direct Wall Mount Double Shelf Bracket
Furniture Shelves
Use the Metro® Super Erecta® Direct Wall Mount Double Shelf Bracket to expand your wall shelving unit.
  • Double shelf brackets are used to create runs of multiple shelves
  • Shelf support brackets can support 14", 18", and 24" wide shelves
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.
Ordering information: A single shelf support (sold separately) is required at the end of each run.