Kinesis® TELOS® neo™ SPE Fixed 96-Well Microplates, Cole-Parmer®
Supplier: Cole-Parmer Instrument Co.
647442 647445 647447 647448 647454 647457 647428 647433 647439
CA76181-368EA 2818.88 CAD
CA76181-368 CA76181-370 CA76181-372 CA76181-374 CA76181-376 CA76181-378 CA76181-428 CA76181-430 CA76181-432
Kinesis® TELOS® neo™ SPE Fixed 96-Well Microplates, Cole-Parmer®
Microplates Solid Phase Extraction Plates (SPE)
Modular design lets you simply remove any unwanted wells and add sealing strips.
  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Polymer based bottom
  • Water-wettable polymer is not affected by drying out
TELOS® neo™ populated plates come with 96 wells already in place ideal for high throughput assays. Polymer-based sorbents have no silanol interactions, excellent flow characteristics, and pH stability providing highly reproducible results. Elution volumes are kept to a minimum byMore Product Information