Kinesis® TELOS® neo™ MicroPlate™ SPE Loose 96-Well Microplates, Cole-Parmer®
Supplier: Cole-Parmer Instrument Co.
647401 647404 647408 647410 647412 647414 647416 647417 647420
CA76181-386PK 2071.85 CAD
CA76181-386 CA76181-392 CA76181-400 CA76181-404 CA76181-408 CA76181-412 CA76181-416 CA76181-418 CA76181-424
Kinesis® TELOS® neo™ MicroPlate™ SPE Loose 96-Well Microplates, Cole-Parmer®
Microplates Solid Phase Extraction Plates (SPE)
Populate base plate with only the wells needed.
  • Elution volume, 50 µl for minimal sample dilution and reduced evaporation time
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Polymer based bottom
The Int.Ø, frit design, and sorbent material of the TELOS® MicroPlate™ loose wells allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µl.
Use the loose wells for assays when the sample numbers vary,More Product Information