DNAOUT™ DNA Removal Solution, G-Biosciences
Supplier: G-Biosciences
786-74
786-75
CA82021-436EA
205.35
CAD
CA82021-436
CA82021-438
DNAOUT™ DNA Removal Solution, G-Biosciences
Cleaning Agents
Decontaminants
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
- Ready-to-use with convenient spray bottle and refill bottles available
- Remove contaminating DNA prior to sensitive work, including PCR
- Removes amplicon, plasmid and genomic DNA
- Fast acting (10-15 seconds)
- Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more