RNaseOUT™, G-Biosciences
Supplier: G-Biosciences
786-70
786-71
CA82021-424EA
196.55
CAD
CA82021-424
CA82021-426
RNaseOUT™, G-Biosciences
Cleaning Agents
Decontaminants
RNaseOUT™ is uniquely formulated to destroy and remove RNases on contact.
- Simply spray and rinse
- RNaseOUT™ is non-toxic and residue free
- RNaseOUT™ allows common equipment to be safely and confidently used for all RNA work