1-Heptanesulfonic acid sodium salt monohydrate ≥99% for HPLC, for ion pair chromatography, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sodium 1-heptanesulfonate monohydrate, Sodium 1-heptanesulphonate monohydrate

HX0101-2 HX0101-3
CAHX0101-2EA 229 CAD
CAHX0101-2 CAHX0101-3
1-Heptanesulfonic acid sodium salt monohydrate ≥99% for HPLC, for ion pair chromatography, Sigma-Aldrich®
1-Heptanesulphonic acid sodium salt monohydrate

Used to separate pharmaceutical compounds from their metabolites


Compounds can be separated on a reversed phase column with the addition of an ion pairing reagent to the mobile phase. The type of ion pairing reagent and its concentration give different selectivities.

Formula: C₇H₁₅NaO₃S·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS Number: 207300-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR