Herculase II Fusion DNA Polymerases, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
600677 600675 600679
CA99902-832EA 345.2 CAD
CA99902-832 CAST600675 CAST600679
Herculase II Fusion DNA Polymerases, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Herculase II Fusion DNA Polymerases produce superior yields with extension times as short as 15 sec/kb, even with genomic DNA targets up to 23kb. Herculase II helps overcome PCR challenges with successful amplification of targets of all complexity. Easily amplify low abundance, high GC, and other difficult targets with robust yields.