Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
BAKR3717-01
BAKR3717-07
CAJT3717-01EA
226.97
CAD
CAJT3717-01
CAJT3717-07
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Sodium hydroxide
Formula:
NaOH Boiling Pt: 1390 °C Melting Pt: 323 °C Density: 2.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets ChP Chemical Specifications | |
Meets E.P. Chemical Specifications | |
Meets J.P. Chemical Specifications | |
Meets B.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Identification A (Sodium) | Passes Test |
NF - Identification B (pH) | ≥11.0 |
NF - Insoluble Substances and Organic Matter | Passes Test |
NF - Potassium (K) | ≤0.5 % |
NF - Carbonate (as Na₂CO₃) | ≤3.0 % |
NF - Total Alkali | 95.0 - 100.5 % |
NF - Content of Sodium | 54.0 - 59.8 % |
ChP -Assay | 97.0 - 100.5 % |
ChP -Sodium carbonate | 2.0% max |
ChP -Identification | Passes Test |
ChP -Alkalinity | Passes Test |
ChP -Clarity and color of solution | Passes Test |
ChP -Chloride | 0.005 % max |
ChP -Sulfate | 0.015 % max |
ChP -Potassium | Passes Test |
ChP -Aluminum and iron | 0.1% max |
ChP -Iron | 0.001% max |
ChP -Heavy metal | 20 ppm max |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Carbonate (as Na₂CO₃) | ≤2.0 % |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Chloride (Cl) | ≤200 ppm |
EP/BP - Sulfate (SO₄) | ≤200 ppm |
EP/BP - Iron (Fe) | ≤10 ppm |
EP/BP - Assay (NaOH) | 97.0 - 100.5 % |
JP - Assay | 95.0 - 101.0 % |
JP - Identification A | Passes Test |
JP - Identification B | Passes Test |
JP - Clarity and Color of Solution | Passes Test |
JP - Chloride (Cl) | ≤0.050 % |
JP - Heavy Metals (as Pb) | ≤30 ppm |
JP - Potassium (K) | Passes Test |
JP - Carbonate (as Na₂CO₃) | ≤2.0 % |
JP - Mercury (Hg) | Passes Test |
Endotoxin Concentration (EU/g) | ≤5 |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...