Ceiling Service Panels, PG LifeLink
Supplier: PG Lifelink
CP24X24CP6S6W CP24X48CP8S6W
76462-644EA 749.57 CAD
76462-644 76462-646
Ceiling Service Panels, PG LifeLink
Furniture Casework
Ceiling Service Panels are ideal for mobile workstations because they create flexibility within a laboratory environment.
  • 16 gauge cold rolled steel with a white power coat finish
  • Six (6) or eight (8) single gang cutouts for power and/or data, each with a junction box (the number of cutouts is based on the panel size)
  • Six (6) 1¼" holes for services (gas, air, vacuum)
  • 1" flange on all sides with (1) hole per side for mounting above the finished ceiling to a permanent support structure
  • Fits most ceiling t-grid systems
Ceiling Service Panels are designed to provide power, phone, data, and/or service outlets (gas, air, vacuum) accessible via umbilical cords, therefore allowing mobile benches to move freely within the lab without the challenge of re-routing services. Ceiling Service Panels can also be used in commercial and retail environments when additional connectivity is needed.
The best method for mounting Ceiling Service Panels will be dictated by field conditions. Mounting hardware to be supplied by electrical contractor or others.
Ordering information: Mounting hardware not included.