Super Erecta® Wall Mount Shelving Post with Brackets, Metro™
Supplier: Metropolitan Wire
13PDFK4 33PDFK4 54PDFK4 63PDFK4
76463-074EA 119.54 CAD
76463-074 76463-100 76463-102 76463-104
Super Erecta® Wall Mount Shelving Post with Brackets, Metro™
Furniture Shelves
Combine with shelves and shelf supports to create a post-type wall mounted shelving solution.
  • 13" posts are ideal for 1-shelf tier configurations
  • 33" posts support 2-shelf configurations
  • 54" posts can accommodate 3 or more shelving tiers
  • 63" posts are designed to handle 4 or more shelving tiers
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.