Buchner Funnels, Table Top, PE, Perforated, 24", Chemglass
Supplier: Chemglass
CG-1413-21 CG-1413-22
16002-746EA 4530.38 CAD
16002-746 16002-748
Buchner Funnels, Table Top, PE, Perforated, 24", Chemglass
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate.
  • Removable hose barb connector accepts 12.7 mm (¹/₂") Int.Ø tube and threads into 3/4" NPT bung on the funnel
  • Fixed or removable perforated filter plates are made of 4.8 mm (³/₁₆") thick HDPE with 4.8 mm (³/₁₆") perforations on 11 mm (⁷/₁₆") centers
  • Use temperature range maximum 52 °C (125 °F), minimum –20 °C (–4 °F)
Perforated filter plates available in two versions (fixed or removable); fixed for coarse filtration or use with a cloth or paper filter; removable for use in precious metals recovery.