Dissecting Forceps
Supplier: DR INSTRUMENTS INC. SE
15C 20L
470315-182BX 625.5 CAD
470315-182 470315-192
Dissecting Forceps
Forceps
Quality fine point forceps.
  • Straight or bent tip
  • Stainless steel construction
  • Textured grip
Dissecting forceps feature straight fine points. Made with high quality stainless steel. Fine points are great for delicate dissection purposes and can be used under the microscope.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.