Hughes Portable, Self-Contained, 10 gal. Gravity-Fed Eyewash Station, Justrite®
Supplier: Justrite
10GFEW 10GFEWP
76462-628EA 428.86 CAD
76462-628 76462-630
Hughes Portable, Self-Contained, 10 gal. Gravity-Fed Eyewash Station, Justrite®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Portable design with carrying handles and concealed wall mounting bracket make moving and relocating easy.
  • Wall mounted
  • High-visibility orange color is easy to identify in emergency
  • Simple to use with pull-down activation handle
  • Wide fill opening enables quick inspection, cleaning, and filling
The Hughes portable, self-contained 10 gal. gravity-fed eyewash station is suitable wherever continuous water isn't available, with no plumbing needed. The gravity-fed eyewash supplies a continuous, controlled flow that exceeds the ANSI Z358.1 standard of 1.5 LPM (0.4 GPM) for 15 minutes, providing a reliable safety solution no matter where it is located. Concealed mounting bracket make moving and relocating easy. High-visibility color is easy to identify in emergency situations. Simple to use with pull-down activation handle.
Mounting type: Wall bracket
Operating time: 15 minutes
Certifications: UL classified to ANSI Z358.1-2014. ANSI Z358.1-2014 and EN 15154 compliant.