SP Wilmad-LabGlass Stem Coaxial Small Volume NMR Inserts, SP Industries
Supplier: SP Industries
WGS-5BL-SP WGS-5BL WGS-10BL
76460-278EA 85.91 CAD
76460-278 76460-276 76460-280
SP Wilmad-LabGlass Stem Coaxial Small Volume NMR Inserts, SP Industries
Tubes NMR Tubes
The most versatile and reliable coaxial system available for NMR experiments.
  • Manufactured from ASTM Type 1 class A glass, ideal for variable temperature studies
  • Outer tube must be ordered separately depending on magnetic field strength
General applications include small volume NMR, external referencing, external locking and magnetic susceptibility determination.