SP Wilmad-LabGlass EPR Cold Finger Liquid Nitrogen Dewars, 50 ml, SP Industries
Supplier: SP Industries
WG-816-B-Q WG-816-Q WG-819-B-Q
76460-246EA 1709.54 CAD
76460-246 76460-248 76460-250 76460-252
SP Wilmad-LabGlass EPR Cold Finger Liquid Nitrogen Dewars, 50 ml, SP Industries
These dewar flasks provide a convenient alternative to VT systems for studies at 77 K.
  • Working time of 15 min after filling
  • Silvered around the reservoir
Offset version designed for cavities with limited space between the reservoir and the drive shaft for iris.
Caution: Check the size of the cavity openings for compatibility.