SP Wilmad-LabGlass Precision Low Pressure Vacuum EPR Tubes, SP Industries
Supplier: SP Industries
727-LPV-250M 734-LPV-7 722-LPV-7
76459-972EA 433.96 CAD
76459-972 76459-962 76459-946
SP Wilmad-LabGlass Precision Low Pressure Vacuum EPR Tubes, SP Industries
Tubes NMR Tubes
A great alternative to J. Young tubes.
  • A 4× larger sealing surface eliminates leaks
Flame-free seal for air sensitive or volatile liquid samples at any pressure from vacuum to pressure slightly above 14.5 psi.
Caution: Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.