SP Wilmad-LabGlass Precision High Pressure Vacuum EPR Tubes, SP Industries
Supplier: SP Industries
700-PV-7 734-PV-7
76459-908EA 790.42 CAD
76459-908 76459-950 76459-976
SP Wilmad-LabGlass Precision High Pressure Vacuum EPR Tubes, SP Industries
Tubes NMR Tubes
This high pressure/vacuum tube is a highly reliable EPR tube for pressure experiments.
  • Valve is made of PTFE and opened by turning counterclockwise
Designed to connect to a ¹/₈" metal (SST or brass) vacuum line using Swagelok® fittings or a rubber vacuum hose.
Caution: Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.