SP Wilmad-LabGlass Low Pressure Vacuum NMR Tubes, SP Industries
Supplier: SP Industries
535-LPV-8 524-LPV-7 528-LPV-8 307-LPV-7 528-LPV-7 522-LPV-7 535-LPV-7 507-LPV-7 528-LPV-7QTZ 507-LPV-8 513-7LPV-7
76460-334EA 291.91 CAD
76460-334 76460-346 76460-332 76460-344 76460-352 76460-330 76460-342 76460-350 76460-338 76460-328 76460-336
SP Wilmad-LabGlass Low Pressure Vacuum NMR Tubes, SP Industries
Tubes NMR Tubes
Ideal for anaerobic and gas-tight NMR experiments, offers a convenient flame-free sealing solution for air sensitive or volatile liquid samples.
  • Robust sealing system allows pressure build-up inside the sample
  • Greaseless PTFE piston provides a 100% contamination-free seal
  • Redesigned with a 4× larger sealing surface; eliminates leaks and greatly increases lifetime when compared to traditional J. Young tubes
  • Axial symmetric design guarantees application in spinning experiments
Caution: Due to the nature of glass, Extreme caution should be exercised when using at elevated or reduced pressures since a tiny scratch on the glass surface would significantly lower the tensile strength. Adequate safety shielding should always be used when working in these conditions.