Gen-X Blue Latex Powder-Free Examination Gloves, Smart Glove
Supplier: VWR International
76449-382 76449-384 76449-386 76449-388 76449-390
76449-382CS 353.86 CAD
76449-382 76449-384 76449-386 76449-388 76449-390
Gen-X Blue Latex Powder-Free Examination Gloves, Smart Glove
Gloves
Gen-X natural rubber latex gloves offer superior comfort to the wearer
  • 5.6 g weight
  • Minimum thickness of 3.1 mil
  • Medical grade
  • Phthalate-free
Natural rubber's elasticity provides a better fit, finger dexterity, excellent tactility and superior sensitivity, which is ideal for minor procedures and examinations.
Certifications: ASTM D3578. Manufactured in compliance with GMP, ISO 9001, ISO 13485, and MDSAP.
Packaging: Packed 100 gloves pers dispenser box, 10 boxes per case.
Caution: This product contains natural rubber latex which may cause allergic reactions. Keep storage area cool, dry, and dust-free, avoid ventilation and storage close to photocopy equipment. Copper ions discolor the glove. Protect gloves against ultraviolet light sources, such as sunlight and oxidizing agents. Storage above 38 °C (100 °F) will lead to accelerated aging and should be avoided under any circumstances.