Ethidium bromide solution 10 mg/mL, Pierce™

Supplier: Thermo Scientific

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide

17898 17898
CAPI17898PK 696.19 CAD
PI17898 CAPI17898
Ethidium bromide solution 10 mg/mL, Pierce™
Ethidium bromide

Thermo Scientific Pierce Ethidium Bromide is available as a dilute and concentrated solution to accommodate most molecular biology applications for fluorescent DNA visualization.


Ethidium Bromide is an intercalating dye used in the visualization of DNA and RNA in electrophoresis gels and cesium chloride density gradients.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
MDL Number: MFCD00011724
CAS Number: 1239-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR