Anti-FMC63 Hybridoma Polyclonal Antibody

Supplier: ACROBIOSYSTEMS INC MS
FM3Y451000TESTS FM3Y45250TESTS FM3Y4525TESTS
103871-626EA 13489.23 CAD
103871-626 103871-628 103871-630
Anti-FMC63 Hybridoma Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FMC63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR