QUANTIFOIL® Multi A Holey Carbon Films on Grids
Supplier: ELECTRON MICROSCOPY SCIENCE
Q2100AMA Q210AMA Q210CMA Q210NMA Q225AMA Q250AMA Q3100AMA Q310AMA Q310CMA Q310NMA Q325AMA Q350AMA Q4100AMA Q410AMA Q410CMA Q410NMA Q425AMA Q450AMA
76440-106PK 2971.54 CAD
76440-106 76440-138 76440-192 76440-246 76440-316 76440-420 76440-522 76440-554 76440-608 76440-662 76440-732 76440-830 76440-928 76440-960 76441-014 76441-068 76441-138 76441-234
QUANTIFOIL® Multi A Holey Carbon Films on Grids
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon request
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement. It has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) microscopy when compared with conventional holey film.
In the Multi A foil the pattern of hole sizes, shapes and arrangements is repeated. In addition to circular holes, the pattern includes oval-shaped ones which appear round at high tilt angles (~70°). The diameters of the round holes are about 1, 1.4, and 2 µm, and the bar widths range from 0.5 to 4 µm. The oval holes in the foil have a dimension of 8×2 µm and 4×1 µm.
The surface properties of QUANTIFOIL holey carbon support film, especially the wetting properties, may have to be adapted according to one's particular requirements. Untreated aging QUANTIFOIL® tends to be hydrophobic. Hydrophillicity of the foil can be achieved by glow discharging in residual air or by metal coating.
Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.