VWR® Microporous Material Lab Coats (Made in USA)
Supplier: VWR International
76457-656 76457-658 76457-660 76457-662 76457-664 76457-666 76457-668 76457-670 76457-672 76457-674 76457-676 76457-678 76457-680 76457-682 76457-684 76457-686 76457-688 76457-690 76457-692 76457-694 76457-696 76457-698 76457-700 76457-702 76470-670 76470-672 76470-674 76470-676 76470-678 76470-680 76470-682 76470-684 76470-686 76470-688 76470-690 76470-692 76470-694 76470-696 76470-698 76470-700
76457-656CS 937.34 CAD
76457-656 76457-658 76457-660 76457-662 76457-664 76457-666 76457-668 76457-670 76457-672 76457-674 76457-676 76457-678 76457-680 76457-682 76457-684 76457-686 76457-688 76457-690 76457-692 76457-694 76457-696 76457-698 76457-700 76457-702 76470-670 76470-672 76470-674 76470-676 76470-678 76470-680 76470-682 76470-684 76470-686 76470-688 76470-690 76470-692 76470-694 76470-696 76470-698 76470-700
VWR® Microporous Material Lab Coats (Made in USA)
Disposable Garments
The VWR® Made in USA PPE line has been expanded to include microporous material laboratory coats.
  • Microporous two-layer material, 60 gsm
  • Fluid and particulate barrier
  • Excellent breathability
  • Serged seams
  • 3 Pocket or no pocket options
  • Knit or standard collar
  • Open, elastic, or knit cuff
Microporous material laboratory coats offer increased fluid protection with great comfort. Available styles include different wrist, collar, and pocket options.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed in cases of 30 coats.