Quantifoil® SiO₂ Holey Carbon Films on Grids
Supplier: Electron Microscopy Sciences
Q225-NS7 Q210AS7 Q210CS7 Q210NS7 Q310AS7 Q310CS7 Q310NS7 Q410AS7 Q410CS7 Q410NS7
100491-236EA 1079.31 CAD
100491-236 76440-118 76440-120 76440-130 76440-132 76440-134 76440-136 76440-186 76440-188 76440-190 76440-240 76440-242 76440-244 76440-286 76440-288 76440-290 76440-354 76440-356 76440-388 76440-390 76440-416 76440-418 76440-456 76440-458 76440-490 76440-492 76440-518 76440-520 76440-534 76440-536 76440-546 76440-548 76440-550 76440-552 76440-602 76440-604 76440-606 76440-656 76440-658 76440-660 76440-702 76440-704 76440-706 76440-766 76440-768 76440-800 76440-802 76440-826 76440-828 76440-864 76440-866 76440-898 76440-900 76440-924 76440-926 76440-940 76440-942 76440-952 76440-954 76440-956 76440-958 76441-008 76441-010 76441-012 76441-062 76441-064 76441-066 76441-108 76441-110 76441-112 76441-170 76441-172 76441-204 76441-206 76441-230 76441-232 76441-266 76441-268 76441-300 76441-302 76441-326 76441-328
Quantifoil® SiO₂ Holey Carbon Films on Grids
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
QUANTIFOIL® with square holes and relatively narrow bars can be used in electron microscopy to support a thin carbon film, which by itself is too fragile to span a grid square. Alternatively, this holey film can directly support an object larger than the holes.
QUANTIFOIL® S 7/2 has a hole size of 7×7 µm. Space between holes is 2 µm. Center to center is 9 µm.
QUANTIFOIL S 35/5 hole size is 35 µm. Space between holes is 5 µm. Center to center is 40 µm.
QUANTIFOIL S 35/10 hole size is 35 µm. Space between holes is 10 µm. Center to center is 45 µm.
Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.