VWR® Advanced Magnetic Hotplate Stirrers
Supplier: VWR International
470330-720 470330-724 470330-732 470330-740 470330-742 470330-744 470330-746 470330-748
470330-720EA 1217.25 CAD
470330-720 470330-724 470330-732 470330-740 470330-742 470330-744 470330-746 470330-748
VWR® Advanced Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
These top-of-the-line units combine advanced digital features with high quality and reliability.
  • All models feature digital displays
  • Speed range: 60 to 1600 rpm (stirring models)
  • Prominent 'Hot' indicator
  • Rocker switch for added safety
  • All models include a 5 year warranty
These high quality units feature advanced digital displays with a prominent hot surface indicator, a rocker switch do disconnect the heater element, audio alarm and overheat protection. Advanced electronics provide high temperature stability (±1%) and speed stability (±2%). The newMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable