SP Wilmad-LabGlass Gas-Tight Septum NMR Tube Caps, SP Industries
Supplier: SP Industries
WG-3889-100 WG-3891-100
76460-192PK 203.91 CAD
76460-192 76460-194
SP Wilmad-LabGlass Gas-Tight Septum NMR Tube Caps, SP Industries
Closures Test/Sample Tube Closures
Gas-tight NMR tube caps are ideal for experiments that require an air-tight environment under vacuum or low pressure less than 1 bar (14.5 psi).
Use these caps with Wilmad medium walled and heavy walled NMR tubes for better seal and robustness in variable temperature experiments. Septa allow easy access via syringe needle.
Caution: Only to be used with Wilmad precision NMR tubes.