SP Wilmad-LabGlass Screw-Cap NMR Tubes, SP Industries
Supplier: SP Industries
335-TR-7 507-TR-7 507-TR-8 513-7TRA-7 528-TR-7 528-TR-7-V17M 528-TR-8 535-TR-8
76459-624EA 89.68 CAD
76459-624 76459-626 76459-628 76459-632 76459-860 76459-862 76459-864 76459-870 76459-872
SP Wilmad-LabGlass Screw-Cap NMR Tubes, SP Industries
Tubes NMR Tubes
These precision screw-cap NMR tubes are manufactured from the highest quality borosilicate glass and are ideal for sample degasification.
  • Maintains a vacuum quality of >10 to 4 torr
  • Ideal for sample degasification
  • Each screw-cap tube comes with one PTFE/silicone septum
PTFE/ rubber septums are inert to most solvents and many corrosive materials but not recommended for multiple punctures.
PTFE/silicone septums are inert to most organic solvents and compounds but not recommended for strongly corrosive materials. The septum remains reliable after multiple punctures.