QUANTIFOIL® R 2 Holey Carbon Films on Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Q225-NR2 Q2100AR1-2NM Q2100AR2-2NM Q2100AR-4-2NM Q2100CR1-2NM Q2100CR2-2NM Q2100CR4-2NM Q210AR1 Q210AR1-2NM Q210AR2 Q210AR2-2NM Q210AR-4 Q210AR-4-2NM Q210CR1 Q210CR1-2NM Q210CR2 Q210CR2-2NM Q210CR-4 Q210CR-4-2NM Q210NR1 Q210NR2 Q210NR-4 Q225AR1-2NM Q225AR2-2NM Q225-AR4 Q225AR4-2NM Q225CR1-2NM Q225CR2-2NM Q225CR4-2NM Q250AR1-2NM Q250AR2-2NM Q250-AR4 Q250AR4-2NM Q250CR1-2NM Q250CR2-2NM Q250CR4-2NM Q3100AR1-2NM Q3100AR2-2NM Q3100AR-4-2NM Q3100CR1-2NM Q3100CR2-2NM Q3100CR-4-2NM Q310AR1 Q310AR1-2NM Q310AR2 Q310AR2-2NM Q310AR-4 Q310AR-4-2NM Q310CR1 Q310CR1-2NM Q310CR2 Q310CR2-2NM Q310CR-4 Q310CR-4-2NM Q310NR1 Q310NR2 Q310NR-4 Q325AR1-2NM Q325AR2-2NM Q325AR-4-2NM Q325CR1-2NM Q325CR2-2NM Q325CR-4-2NM Q350AR1-2NM Q350AR2-2NM Q350AR-4-2NM Q350CR1-2NM Q350CR2-2NM Q350CR-4-2NM Q4100AR1-2NM Q4100AR2-2NM Q4100AR-4-2NM Q4100CR1-2NM Q4100CR2-2NM Q4100CR-4-2NM Q410AR1 Q410AR1-2NM Q410AR2 Q410AR2-2NM Q410AR-4 Q410AR-4-2NM Q410CR1 Q410CR1-2NM Q410CR2 Q410CR2-2NM Q410CR-4 Q410CR-4-2NM Q410NR1 Q410NR2 Q410NR-4 Q425AR1-2NM Q425AR2-2NM Q425AR-4-2NM Q425CR1-2NM Q425CR2-2NM Q425CR-4-2NM Q450AR1-2NM Q450AR2-2NM Q450AR-4-2NM Q450CR1-2NM Q450CR2-2NM Q450CR-4-2NM
100491-232EA 883.08 CAD
100491-232 76440-112 76440-114 76440-116 76440-124 76440-126 76440-128 76440-142 76440-156 76440-162 76440-166 76440-172 76440-174 76440-196 76440-210 76440-216 76440-220 76440-226 76440-228 76440-250 76440-266 76440-274 76440-330 76440-336 76440-340 76440-342 76440-368 76440-374 76440-378 76440-432 76440-438 76440-442 76440-444 76440-470 76440-476 76440-480 76440-528 76440-530 76440-532 76440-540 76440-542 76440-544 76440-558 76440-572 76440-578 76440-582 76440-588 76440-590 76440-612 76440-626 76440-632 76440-636 76440-642 76440-644 76440-666 76440-682 76440-690 76440-746 76440-752 76440-756 76440-780 76440-786 76440-790 76440-844 76440-850 76440-854 76440-878 76440-884 76440-888 76440-934 76440-936 76440-938 76440-946 76440-948 76440-950 76440-964 76440-978 76440-984 76440-988 76440-994 76440-996 76441-018 76441-032 76441-038 76441-042 76441-048 76441-050 76441-072 76441-088 76441-096 76441-150 76441-156 76441-160 76441-184 76441-190 76441-194 76441-246 76441-252 76441-256 76441-280 76441-286 76441-290
QUANTIFOIL® R 2 Holey Carbon Films on Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon request
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL® with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness.
The hole size chosen depends on the magnification used, and on whether or not one wishes to include support film in the image. Assessment of the image quality is easier when foil is included in the picture, because the power spectrum of a foil is stronger than that of unsupported ice.
QUANTIFOIL® R 2/1 has more open area than R 2/2. The hole size is 2 µm. Space between holes is 1 µm. It is used when focusing is carried out on the edge of a hole burnt in the ice in a neighboring hole instead of on the foil adjacent to the hole.
QUANTIFOIL® R 2/2 has a hole size of 2 µm. Space between holes is 2 µm. Center to center is 4 µm. This type is used at magnifications between 30000x and 40000x.
QUANTIFOIL® R 2/4 has a hole size of 2 µm. Space between holes is 4 µm. Center to center is 6 µm. This type may be preferred over R 2/2, when an increased tolerance with respect to the position of beam, and a larger beam Ø are desired, such as in the case of automated image acquisition.
Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.