PTB7-Th (PCE10), Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66975-MC
H66975-MD
H66975-ME
AAH66975-MCEA
1004.88
CAD
AAH66975-MC
AAH66975-MD
AAH66975-ME
PTB7-Th (PCE10), Organic Photovoltaics (OPV) reagent
PTB7-Th (PCE10)
Formula:
(C₄₉H₅₇FO₂S₆)n |
CAS Number:
1469791-66-9 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.58 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...