IEICO-4Cl, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66752-MC
H66752-MD
H66752-ME
AAH66752-MCEA
835.82
CAD
AAH66752-MC
AAH66752-MD
AAH66752-ME
IEICO-4Cl, Organic Photovoltaics (OPV) reagent
IEICO-4Cl
Formula:
C₁₁₄H₁₁₄Cl₄N₄O₄S₄ |
CAS Number:
2240998-88-1 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.26 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...