PDCBT, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66713-MC
H66713-MD
H66713-ME
AAH66713-MCEA
922.72
CAD
AAH66713-MC
AAH66713-MD
AAH66713-ME
PDCBT, Organic Photovoltaics (OPV) reagent
PDCBT
Formula:
(C₄₄H₆₂O₄S₄)n |
CAS Number:
1609536-17-5 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.89 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...