ITIC-F, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66664-MC
H66664-MD
H66664-ME
AAH66664-MCEA
837.4
CAD
AAH66664-MC
AAH66664-MD
AAH66664-ME
ITIC-F, Organic Photovoltaics (OPV) reagent
ITIC-F
Formula:
C₉₄H₇₈F₄N₄O₂S₄ MW: 1499.88 g/mol |
CAS Number:
2097998-59-7 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.55 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...