Y5, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66460-MC
H66460-MD
H66460-ME
AAH66460-MCEA
922.72
CAD
AAH66460-MC
AAH66460-MD
AAH66460-ME
Y5, Organic Photovoltaics (OPV) reagent
Y5
Formula:
C₈₂H₉₀N₈O₂S₅ |
CAS Number:
2304444-48-0 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.41 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...