IDT-2BR, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66830-MC
H66830-MD
H66830-ME
AAH66830-MCEA
862.68
CAD
AAH66830-MC
AAH66830-MD
AAH66830-ME
IDT-2BR, Organic Photovoltaics (OPV) reagent
IDT-2BR
Formula:
C₈₈H₈₈N₆O₂S₈ MW: 1518.20 g/mol |
CAS Number:
2042521-91-3 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.69 ± 0.02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...