IEICO, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66656-MC
H66656-MD
H66656-ME
AAH66656-MCEA
861.1
CAD
AAH66656-MC
AAH66656-MD
AAH66656-ME
IEICO, Organic Photovoltaics (OPV) reagent
IEICO
Formula:
C₁₁₄H₁₁₈N₄O₄S₄ |
CAS Number:
2055812-53-6 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.34 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...