ITIC, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66574-MC
H66574-MD
H66574-ME
AAH66574-MCEA
671.5
CAD
AAH66574-MC
AAH66574-MD
AAH66574-ME
ITIC, Organic Photovoltaics (OPV) reagent
ITIC
Formula:
C₉₄H₈₂N₄O₂S₄ MW: 1427.94 g/mol |
CAS Number:
1664293-06-4 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.57 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...