IEICO-4F, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66546-MC
H66546-MD
H66546-ME
AAH66546-MCEA
835.82
CAD
AAH66546-MC
AAH66546-MD
AAH66546-ME
IEICO-4F, Organic Photovoltaics (OPV) reagent
IEICO-4F
Formula:
C₁₁₄H₁₁₄F₄N₄O₄S₄ |
CAS Number:
2089044-02-8 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.28 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...