PBDB-T (PCE12), OPV - Organic Photovoltaics
Supplier: Thermo Scientific Chemicals
H66526-MC
H66526-MD
H66526-ME
AAH66526-MCEA
671.5
CAD
AAH66526-MC
AAH66526-MD
AAH66526-ME
PBDB-T (PCE12), OPV - Organic Photovoltaics
PBDB-T (PCE12)
Formula:
(C₆₈H₇₈O₂S₈)n |
CAS Number:
1415929-80-4 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.82 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...