ITIC-Cl, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific Chemicals

H66521-MC H66521-MD H66521-ME
AAH66521-MCEA 837.4 CAD
AAH66521-MC AAH66521-MD AAH66521-ME
ITIC-Cl, Organic Photovoltaics (OPV) reagent
ITIC-Cl
Formula: C₉₄H₇₈Cl₄O₂S₄
MW: 1565.72 g/mol
CAS Number: 2253663-81-7

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.46 ± 0.02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR