PDPPTT, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly[2,5-(2-decyltetradecyl)-3,6-diketopyrrolopyrrole-alt-5,5-(2,5- di(thien-2- yl)thieno[3,2-b]thiophene)]
H66319-MC
H66319-MD
H66319-ME
AAH66319-MCEA
835.82
CAD
AAH66319-MC
AAH66319-MD
AAH66319-ME
PDPPTT, Organic Photovoltaics (OPV) reagent
PDPPTT
Formula:
(C₆₈H₁₀₄N₂O₂S₄)n |
CAS Number:
1260685-66-2 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.40 ± 0.02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...