BTP-4Cl, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
TTPTTI-4C
H66315-MC
H66315-MD
H66315-ME
AAH66315-MCEA
921.14
CAD
AAH66315-MC
AAH66315-MD
AAH66315-ME
BTP-4Cl, Organic Photovoltaics (OPV) reagent
BTP-4Cl
Formula:
C₈₂H₈₆Cl₄N₈O₂S₅ |
CAS Number:
2414918-25-3 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.36 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...