PBDB-T-2F (PCE14), Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
PCE135
H66179-MC
H66179-MD
H66179-ME
AAH66179-MCEA
921.14
CAD
AAH66179-MC
AAH66179-MD
AAH66179-ME
PBDB-T-2F (PCE14), Organic Photovoltaics (OPV) reagent
PBDB-T-2F (PCE14)
Formula:
(C₆₈H₇₆F₂O₂S₈)n |
CAS Number:
1802013-83-7 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.85 ± 0.02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...