EH-IDTBr, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific Chemicals

Synonyms: 5,5'-(((4,4,9,9-Tetrakis(2-ethylhexyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(benzo[c][1,2,5]thiadiazole-7,4-diyl))bis(methaneylylidene))bis(3-ethyl-2-thioxothiazolidin-4-one)

H66142-MD H66142-ME H66142-MC
AAH66142-MEEA 3009.9 CAD
AAH66142-ME AAH66142-MD AAH66142-MC
EH-IDTBr, Organic Photovoltaics (OPV) reagent
EH-IDTBr
Formula: C₇₂H₈₈N₆O₂S₈
MW: 1326.03 g/mol
CAS Number: 2102510-60-9

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.64 ± 0.02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR